Sodium dodecyl sulfate (SDS) ≥97.0% (by GC, titration analysis)
Supplier: TCI America
|
Danger
|
CAS Number: 151-21-3
MDL Number: MFCD00036175
Molecular Formula: C12H26O4S
Molecular Weight: 288.38
Purity/Analysis Method:
Product Categories: Materials Chemistry, Anionic Surfactants
Other Product Names / Synonyms: Dodecylsulfuric Acid Sodium Salt, Laurylsulfuric Acid Sodium Salt, Sodium Lauryl Sulfate, SDS
Formula:
CH₃(CH₂)₁₁OSO₃Na Melting Pt: 207 °C Density: 1.1 g/cm³ (20 °C) Flash Pt: 10 °C Storage Temperature: Ambient |
MDL Number:
MFCD00036175 CAS Number: 151-21-3 EINECS: 205-788-1 UN: 1325 ADR: 4.1,II Merck Index: 14,08636 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...