Sodium dodecyl sulfate (SDS) ≥97.0%

Supplier: TCI America
Danger

Synonyms: Sodium lauryl sulfate, Dodecyl sulfate sodium salt, Lauryl sulfate sodium salt, Dodecyl sulfuric acid sodium salt, Dodecyl sulphate sodium salt, Sodium dodecyl sulfate, Dodecyl sulphuric acid sodium salt, Lauryl sulphate sodium salt, Sodium lauryl sulphate

D1403-25G D1403-500G
TCD1403-025GEA 16.28 USD
TCD1403-025G TCD1403-500G
Sodium dodecyl sulfate (SDS) ≥97.0%
Sodium dodecyl sulphate (SDS)

CAS Number: 151-21-3 MDL Number: MFCD00036175 Molecular Formula: C12H26O4S Molecular Weight: 288.38 Purity/Analysis Method: 97.0% (GC,T) Form: Crystal Melting point (°C): 207 Flash Point (°C): 10


Product Categories: Materials Chemistry, Anionic Surfactants Other Product Names / Synonyms: Dodecylsulfuric Acid Sodium Salt, Laurylsulfuric Acid Sodium Salt, Sodium Lauryl Sulfate, SDS

Formula: CH₃(CH₂)₁₁OSO₃Na
Melting Pt: 207 °C
Density: 1.1 g/cm³ (20 °C)
Flash Pt: 10 °C
Storage Temperature: Ambient
MDL Number: MFCD00036175
CAS Number: 151-21-3
EINECS: 205-788-1
UN: 1325
ADR: 4.1,II
Merck Index: 14,08636
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=38f0929c97ab1c86d41ac28c906e7b67b678873f&vid=071d621c42ae42a23801f880f3a9fed48ab461e3&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR