Dibutyl disulfide ≥98.5%

Supplier: TCI America
Warning

Synonyms: Dibutyldisulphide, n-Butyl disulfide, n-Butyl disulphide, Dibutyldisulfide

D1395-25ML
TCD1395-025MLEA 52 USD
TCD1395-025ML
Dibutyl disulfide ≥98.5%
Dibutyl disulphide

[for Determination of Sulfur Content by Energy Dispersive X-ray Fluorescence Method] CAS Number: 629-45-8 MDL Number: MFCD00009467 Molecular Formula: C8H18S2 Molecular Weight: 178.35 Purity/Analysis Method: 98.5% (GC) Form: Clear Liquid Boiling point (°C): 226 Flash Point (°C): 93 Specific Gravity (20/20): 0.94


Other Product Names / Synonyms: Butyl Disulfide, DBDS

Formula: C₈H₁₈S₂
MW: 178.36 g/mol
Boiling Pt: 226 °C
Melting Pt: –94 °C
Density: 0.94 (20 °C)
Flash Pt: 93 °C
Storage Temperature: Ambient
MDL Number: MFCD00009467
CAS Number: 629-45-8
EINECS: 211-091-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=230ef0afc5dc314323697d738f50a99d9a541e39&vid=230ef0afc5dc314323697d738f50a99d9a541e39&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.5% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR