Dabsyl chloride ≥98.0% (by titrimetric analysis)
Supplier: TCI America
[N-Protecting Agent for Peptides Research]
CAS Number: 56512-49-3
MDL Number: MFCD00007444
Molecular Formula: C14H14ClN3O2S
Molecular Weight: 323.80
Purity/Analysis Method:
Product Categories: Analytical Science, UV Detection (HPLC Labeling Reagents)
Other Product Names / Synonyms: 4-Dimethylaminoazobenzene-4'-sulfonyl Chloride
Formula:
C₁₄H₁₄ClN₃O₂S |
MDL Number:
MFCD00007444 CAS Number: 56512-49-3 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...