2,3-Dimethylhexane ≥94.0%

Supplier: TCI America
Danger

D1225-5ML
TCD1225-005MLEA 365 USD
TCD1225-005ML
2,3-Dimethylhexane ≥94.0%
2,3-Dimethylhexane

CAS Number: 584-94-1 MDL Number: MFCD00039951 Molecular Formula: C8H18 Molecular Weight: 114.23 Purity/Analysis Method: 94.0% (GC) Form: Clear Liquid Boiling point (°C): 115 Flash Point (°C): 7 Specific Gravity (20/20): 0.71

Formula: C₈H₁₈
Boiling Pt: 115 °C
Melting Pt: –124 °C
Density: 0.71 (20 °C)
Flash Pt: 7 °C
Storage Temperature: Ambient
MDL Number: MFCD00039951
CAS Number: 584-94-1
EINECS: 209-547-1
UN: 1262
ADR: 3,II

Order Now

Specification Test Results

Purity/Analysis Method 94.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR