Dipropyl sulfone ≥99.0%

Supplier: TCI America

D1171-25G
TCD1171-025GEA 137 USD
TCD1171-025G
Dipropyl sulfone ≥99.0%
Dipropyl sulphone

CAS Number: 598-03-8 MDL Number: MFCD00007570 Molecular Formula: C6H14O2S Molecular Weight: 150.24 Purity/Analysis Method: 99.0% (GC) Form: Crystal Flash Point (°C): 126 Freezing point (°C): 31


Product Categories: Materials Chemistry, Secondary Battery Research Reagents Other Product Names / Synonyms: Propyl Sulfone

Formula: C₆H₁₄O₂S
Boiling Pt: 70…73 °C (0.3 mmHg)
Melting Pt: 29.5 °C
Flash Pt: 126 °C
MDL Number: MFCD00007570
CAS Number: 598-03-8
EINECS: 209-913-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c36a64b1ce149c691366e9ca54f6e955ec89aefd&vid=c36a64b1ce149c691366e9ca54f6e955ec89aefd&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR