Chloro(dimethyl)(phenyl)silane ≥96.0% (by GC)

Supplier: TCI America

Synonyms: Chlorodimethylphenylsilane

D1147-25ML
TCD1147-25MLEA 74 USD
TCD1147-25ML
Chloro(dimethyl)(phenyl)silane ≥96.0% (by GC)
Chloro(dimethyl)(phenyl)silane

CAS Number: 768-33-2 MDL Number: MFCD00000499 Molecular Formula: C8H11ClSi Molecular Weight: 170.71 Purity/Analysis Method: 96.0% (GC) Form: Clear Liquid Boiling point (°C): 198 Flash Point (°C): 82 Specific Gravity (20/20): 1.03


Product Categories: Synthetic Organic Chemistry, Silicon Compounds (for Synthesis), Materials Chemistry, Silane Coupling Agents Other Product Names / Synonyms: Phenyldimethylchlorosilane

Formula: C₈H₁₁ClSi
MW: 170.71 g/mol
Boiling Pt: 198 °C
Density: 1.03 (20 °C)
Flash Pt: 82 °C
MDL Number: MFCD00000499
CAS Number: 768-33-2
UN: 2987
ADR: 8,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b1268fe2b596dd5437377e191d895325c98c7b3c&vid=b1268fe2b596dd5437377e191d895325c98c7b3c&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 96.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR