Diethylaminoacetonitrile ≥98.0% (by GC, titration analysis)

Supplier: TCI America

D1104-25ML
TCD1104-025MLEA 145.48 USD
TCD1104-025ML
Diethylaminoacetonitrile ≥98.0% (by GC, titration analysis)
Diethylaminoacetonitrile

CAS Number: 3010-02-4 MDL Number: MFCD00001891 Molecular Formula: C6H12N2 Molecular Weight: 112.18 Purity/Analysis Method: 98.0% (GC,T) Form: Clear Liquid Boiling point (°C): 170 Flash Point (°C): 53 Specific Gravity (20/20): 0.87


Other Product Names / Synonyms: N,N-Diethylglycinonitrile

Formula: C₆H₁₂N₂
Boiling Pt: 170 °C
Density: 0.87 (20 °C)
Flash Pt: 53 °C
MDL Number: MFCD00001891
CAS Number: 3010-02-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a7763c3b6937d6d8e7960c340d1fb84c6904ab93&vid=a7763c3b6937d6d8e7960c340d1fb84c6904ab93&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR