cis-1,3-Dimethylcyclohexane ≥99.0%

Supplier: TCI America

D1055-25ML
TCD1055-025MLEA 221 USD
TCD1055-025ML
cis-1,3-Dimethylcyclohexane ≥99.0%
cis-1,3-Dimethylcyclohexane

CAS Number: 638-04-0 MDL Number: MFCD00001507 Molecular Formula: C8H16 Molecular Weight: 112.22 Purity/Analysis Method: 99.0% (GC) Form: Clear Liquid Boiling point (°C): 120 Flash Point (°C): 6 Specific Gravity (20/20): 0.77


Other Product Names / Synonyms: cis-Hexahydro-m-xylene

Formula: C₈H₁₆
Boiling Pt: 120 °C
Melting Pt: –75.5 °C
Density: 0.77 (20 °C)
Flash Pt: 6 °C
MDL Number: MFCD00001507
CAS Number: 638-04-0
EINECS: 211-316-5
UN: 2263
ADR: 3,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ab36b3d01e48f9120dec9e5779768ae17a4ffc12&vid=ab36b3d01e48f9120dec9e5779768ae17a4ffc12&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR