Dipropyleneglycol monomethyl ether (mixture of isomers) ≥98.0%

Supplier: TCI America

D1053-25G D1053-500G
TCD1053-025GEA 18 USD
TCD1053-025G TCD1053-500G
Dipropyleneglycol monomethyl ether (mixture of isomers) ≥98.0%
Dipropyleneglycol monomethyl ether (mixture of isomers)

CAS Number: 34590-94-8 MDL Number: MFCD00059604 Molecular Formula: C7H16O3 Molecular Weight: 148.20 Purity/Analysis Method: 98.0% (GC) Form: Clear Liquid Boiling point (°C): 188 Melting point (°C): -80 Flash Point (°C): 77 Specific Gravity (20/20): 0.95

Formula: C₇H₁₆O₃
Boiling Pt: 188 °C
Melting Pt: –80 °C
Density: 0.95 (20 °C)
Flash Pt: 77 °C
MDL Number: MFCD00059604
CAS Number: 34590-94-8
EINECS: 252-104-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8f903db8ad2670c203820d1bff6a2bf10b568540&vid=8f903db8ad2670c203820d1bff6a2bf10b568540&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR