Dibenzo[b,def]chrysene ≥98.0%

Supplier: TCI America

Synonyms: 3,4:8,9-Dibenzpyrene, Dibenzo[a,h]pyrene

D1005-100MG
TCD1005-100MGEA 450 USD
TCD1005-100MG
Dibenzo[b,def]chrysene ≥98.0%
Dibenzo[b,def]chrysene

CAS Number: 189-64-0 MDL Number: MFCD00039579 Molecular Formula: C24H14 Molecular Weight: 302.38 Purity/Analysis Method: 98.0% (GC) Form: Crystal Boiling point (°C): 596 Melting point (°C): 308 Flash Point (°C): -11


Product Categories: Materials Chemistry, Nanocarbon Unit Structures (Nanocarbon Materials) Other Product Names / Synonyms: Dibenzo[a,h]pyrene, 3,4:8,9-Dibenzpyrene

Formula: C₂₄H₁₄
MW: 302.38 g/mol
Boiling Pt: 596 °C
Melting Pt: 308 °C
Flash Pt: –11 °C
MDL Number: MFCD00039579
CAS Number: 189-64-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=aa429b34a82a94c19805115c5cdfac7b1d308494&vid=aa429b34a82a94c19805115c5cdfac7b1d308494&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR