n-Dotriacontane ≥96.0%

Supplier: TCI America

Synonyms: Dotriacontane

D0998-25G D0998-5G
TCD0998-25GEA 225.89 USD
TCD0998-25G TCD0998-005G
n-Dotriacontane ≥96.0%
n-Dotriacontane

CAS Number: 544-85-4 MDL Number: MFCD00009411 Molecular Formula: C32H66 Molecular Weight: 450.88 Purity/Analysis Method: 96.0% (GC) Form: Crystal Boiling point (°C): 220 Melting point (°C): 70 Flash Point (°C): 243

Formula: H₃C(CH₂)₃₀CH₃
Boiling Pt: 220 °C
Melting Pt: 70 °C
Density: 0.7798 g/cm³ (20 °C)
Flash Pt: 243 °C
Storage Temperature: Ambient
MDL Number: MFCD00009411
CAS Number: 544-85-4
EINECS: 208-881-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=48620eec7b709003ba44122386606e277cae14f7&vid=00045f5790b175924aebd285cc3080d00f11bf6b&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 96.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR