Sodium dodecyl sulfate (SDS) ≥85.0%

Supplier: TCI America
Danger

Synonyms: Sodium lauryl sulfate, Dodecyl sulfate sodium salt, Lauryl sulfate sodium salt, Dodecyl sulfuric acid sodium salt, Dodecyl sulphate sodium salt, Sodium dodecyl sulfate, Dodecyl sulphuric acid sodium salt, Lauryl sulphate sodium salt, Sodium lauryl sulphate

D0996-500G D0996-25G
TCD0996-500GEA 50.12 USD
TCD0996-500G TCD0996-025G
Sodium dodecyl sulfate (SDS) ≥85.0%
Sodium dodecyl sulphate (SDS)

CAS Number: 151-21-3 MDL Number: MFCD00036175 Molecular Formula: C12H26O4S Molecular Weight: 288.38 Purity/Analysis Method: 85.0% (T) Form: Crystal Melting point (°C): 207


Product Categories: Materials Chemistry, Anionic Surfactants Other Product Names / Synonyms: Dodecylsulfuric Acid Sodium Salt, Laurylsulfuric Acid Sodium Salt, Sodium Lauryl Sulfate, SDS

Formula: CH₃(CH₂)₁₁OSO₃Na
Melting Pt: 207 °C
Density: 1.1 g/cm³ (20 °C)
Flash Pt: >100 °C
Storage Temperature: Ambient
MDL Number: MFCD00036175
CAS Number: 151-21-3
EINECS: 205-788-1
UN: 1325
ADR: 4.1,II
Merck Index: 14,08636
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3af268a7380d51ba135b686612a8042cf953b348&vid=6ad5f4c94cb0bfebeeb0ae8c15566e9272a07b2d&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 85.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR