Sodium dodecylbenzenesulfonate ≥95.0% (by titrimetric analysis), hard type

Supplier: TCI America
Danger

D0990-25G
TCD0990-025GEA 30.59 USD
TCD0990-025G
Sodium dodecylbenzenesulfonate ≥95.0% (by titrimetric analysis), hard type
Sodium dodecylbenzenesulphonate

CAS Number: 25155-30-0 MDL Number: MFCD00011508 Molecular Formula: C18H30O3S Molecular Weight: 348.48 Purity/Analysis Method: 95.0% (T) Form: Crystal


Product Categories: Materials Chemistry, Anionic Surfactants Other Product Names / Synonyms: Alkylbenzenesulfonic Acid Sodium Salt, Dodecylbenzenesulfonic Acid Sodium Salt, Laurylbenzenesulfonic Acid Sodium Salt, Sodium Alkylbenzenesulfonate, Sodium Laurylbenzenesulfonate

Formula: C₁₈H₂₉NaO₃S
Melting Pt: > 300 °C
Density: 1 g/cm³ (20 °C)
MDL Number: MFCD00011508
CAS Number: 25155-30-0
EINECS: 246-680-4
Merck Index: 13,08686

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR