2,6-Dihydroxynaphthalene ≥95.0%
Supplier: TCI America
|
Warning
|
Synonyms:
Naphthalene-2,6-diol
D0956-5G
D0956-25G
TCD0956-005GEA
53
USD
TCD0956-005G
TCD0956-25G
2,6-Dihydroxynaphthalene ≥95.0%
2,6-Dihydroxynaphthalene
CAS Number: 581-43-1
MDL Number: MFCD00004082
Molecular Formula: C10H8O2
Molecular Weight: 160.17
Purity/Analysis Method:
Product Categories: Materials Chemistry, Monomers
Other Product Names / Synonyms: 2,6-Naphthalenediol
Formula:
C₁₀H₈O₂ MW: 160.17 g/mol Boiling Pt: 375 Melting Pt: 220 Density: 1.33 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00004082 CAS Number: 581-43-1 EINECS: 209-465-6 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...