meso-ɑ,β-Di(4-pyridyl) glycol ≥98.0% (by HPLC, titration analysis)
Supplier: TCI America
CAS Number: 4972-49-0
MDL Number: MFCD00023627
Molecular Formula: C12H12N2O2
Molecular Weight: 216.24
Purity/Analysis Method:
Other Product Names / Synonyms: meso-1,2-Bis(4-pyridyl) Glycol, meso-1,2-Di(4-pyridyl) Glycol, meso-1,2-Di(4-pyridyl)-1,2-ethanediol
Formula:
C₁₂H₁₂N₂O₂ |
MDL Number:
MFCD00023627 CAS Number: 4972-49-0 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...