N,N-Diphenylcarbamoyl chloride ≥98.0% (by GC, titration analysis)

Supplier: TCI America

D0880-25G
TCD0880-025GEA 165 USD
TCD0880-025G
N,N-Diphenylcarbamoyl chloride ≥98.0% (by GC, titration analysis)
N,N-Diphenylcarbamoyl chloride

CAS Number: 83-01-2 MDL Number: MFCD00000633 Molecular Formula: C13H10ClNO Molecular Weight: 231.68 Purity/Analysis Method: 98.0% (GC,T) Form: Crystal Melting point (°C): 83

Formula: C₁₃H₁₀ClNO
Melting Pt: 83 °C
MDL Number: MFCD00000633
CAS Number: 83-01-2
EINECS: 201-450-2
UN: 3261
ADR: 8,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f9fc47ae4740838ba188203ee51b6727b1afa406&vid=f9fc47ae4740838ba188203ee51b6727b1afa406&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR