Citral ≥98.0%

Supplier: TCI America
Warning

Synonyms: 3,7-Dimethylocta-2,6-dienal

D0762-25ML
TCD0762-025MLEA 27.61 USD
TCD0762-025ML
Citral ≥98.0%
Citral

CAS Number: 5392-40-5 MDL Number: MFCD00006997 Molecular Formula: C10H16O Molecular Weight: 152.24 Purity/Analysis Method: 98.0% (GC) Form: Clear Liquid Boiling point (°C): 229 Flash Point (°C): 104 Specific Gravity (20/20): 0.89


Product Categories: Bioscience, Crude Medicine Ingredients for Research and Experimental Use, Terpenoids (Phytochemicals), Acyclic Monoterpenes Other Product Names / Synonyms: 3,7-Dimethyl-2,6-octadienal

Formula: C₁₀H₁₆O
MW: 152.24 g/mol
Boiling Pt: 229 °C
Melting Pt: < –10 °C
Density: 0.89 (20 °C)
Flash Pt: 104 °C
Storage Temperature: Ambient
MDL Number: MFCD00006997
CAS Number: 5392-40-5
EINECS: 226-394-6
Merck Index: 13,02346
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=28b19d63ba7ba7dec451df9584c6af72691d80c8&vid=7b43845d29619fd5a36dc620f0bff17409570d0e&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR