Bathocuproin ≥98.0%
Supplier: TCI America
Synonyms:
2,9-Dimethyl-4,7-diphenyl-1,10-phenanthroline
D0711-5G
D0711-1G
TCD0711-005GEA
395.4
USD
TCD0711-005G
TCD0711-001G
Bathocuproin ≥98.0%
Bathocuproin
CAS Number: 4733-39-5
MDL Number: MFCD00004972
Molecular Formula: C26H20N2
Molecular Weight: 360.46
Purity/Analysis Method:
Product Categories: Synthetic Organic Chemistry, Ligands, Materials Chemistry, Organic Light-Emitting Diode (OLED) Materials, Analytical Science, Phenanthrolines (Chelating Agents)
Other Product Names / Synonyms: 2,9-Dimethyl-4,7-diphenyl-1,10-phenanthroline
Formula:
C₂₆H₂₀N₂ Melting Pt: 283 °C |
MDL Number:
MFCD00004972 CAS Number: 4733-39-5 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...