2,3-Dimethyl-2-butene ≥96.0%

Supplier: TCI America

Synonyms: Tetramethylethylene

D0693-25ML D0693-500ML
TCD0693-025MLEA 79 USD
TCD0693-025ML TCD0693-500ML
2,3-Dimethyl-2-butene ≥96.0%
2,3-Dimethyl-2-butene

CAS Number: 563-79-1 MDL Number: MFCD00008897 Molecular Formula: C6H12 Molecular Weight: 84.16 Purity/Analysis Method: 96.0% (GC) Form: Clear Liquid Boiling point (°C): 73 Flash Point (°C): -16 Specific Gravity (20/20): 0.71


Other Product Names / Synonyms: Tetramethylethylene

Formula: C₆H₁₂
MW: 84.16 g/mol
Boiling Pt: 73
Melting Pt: -75
Density: 0.71 (20 °C)
Flash Pt: –16 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00008897
CAS Number: 563-79-1
EINECS: 209-263-8
UN: 3295
ADR: 3 II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8f247171a650f73b84e1e8593af7d584eeff24af&vid=ae90bd0b11a04136f21e4f2f0d25bdc9e36065a9&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 96.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR