Dimethyl acetal ≥98.0%

Supplier: TCI America
Danger

D0633-500ML D0633-25ML
TCD0633-500MLEA 501 USD
TCD0633-500ML TCD0633-25ML
Dimethyl acetal ≥98.0%
Dimethyl acetal

CAS Number: 534-15-6 MDL Number: MFCD00008493 Molecular Formula: C4H10O2 Molecular Weight: 90.12 Purity/Analysis Method: 98.0% (GC) Form: Clear Liquid Boiling point (°C): 65 Melting point (°C): -113 Flash Point (°C): 1 Specific Gravity (20/20): 0.85


Other Product Names / Synonyms: Acetaldehyde Dimethyl Acetal, 1,1-Dimethoxyethane, Ethylidene Dimethyl Ether

Formula: C₄H₁₀O₂
Boiling Pt: 65 °C
Melting Pt: –113 °C
Density: 0.85 (20 °C)
Flash Pt: 1 °C
MDL Number: MFCD00008493
CAS Number: 534-15-6
EINECS: 208-589-8
UN: 2377
ADR: 3,II
Merck Index: 13,03253
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4dadeb4178514c7fdde10ec72d9d984328d2214d&vid=4dadeb4178514c7fdde10ec72d9d984328d2214d&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR