3-(3,4-Dihydroxyphenyl)-L-alanine ≥98.0%

Supplier: TCI America

Synonyms: Dopar, L-dopa, L-β-3,4-Dihydroxyphenyl-Alanine, 3-Hydroxy-L-tyrosine, Levodopa

D0600-5G D0600-25G D0600-100G
TCD0600-005GEA 48.7 USD
TCD0600-005G TCD0600-25G TCD0600-100G
3-(3,4-Dihydroxyphenyl)-L-alanine ≥98.0%
3-(3,4-Dihydroxyphenyl)-L-alanine

CAS Number: 59-92-7 MDL Number: MFCD00002598 Molecular Formula: C9H11NO4 Molecular Weight: 197.19 Purity/Analysis Method: 98.0% (HPLC,T) Form: Crystal Melting point (°C): 278 Specific rotation [a]20/D: -12 deg (C=5, 1mol/L HCl)


Product Categories: Bioscience, Stains & Dyes, Neurotransmitter Agent Ingredients, Prodrug Ingredients, Biologically-modified Amino Acids Other Product Names / Synonyms: L-DOPA, Levodopa

Formula: C₉H₁₁NO₄
MW: 197.19 g/mol
Melting Pt: 278 °C
MDL Number: MFCD00002598
CAS Number: 59-92-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ca0c0cac1831886d7c48a94772f95dcc4fca0784&vid=ca0c0cac1831886d7c48a94772f95dcc4fca0784&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR