3-(3,4-Dihydroxyphenyl)-DL-alanine ≥98.0%
Supplier: TCI America
Synonyms:
DL-β-3,4-Dihydroxyphenyl-Alanine, DL-DOPA, 3,4-Dihydroxy-DL-phenylalanine
D0599-1G
TCD0599-001GEA
48
USD
TCD0599-001G
3-(3,4-Dihydroxyphenyl)-DL-alanine ≥98.0%
3-(3,4-Dihydroxyphenyl)-DL-alanine
CAS Number: 63-84-3
MDL Number: MFCD00063060
Molecular Formula: C9H11NO4
Molecular Weight: 197.19
Purity/Analysis Method:
Product Categories: Bioscience, Biologically-modified Amino Acids
Other Product Names / Synonyms: DL-DOPA
Formula:
C₉H₁₁NO₄ Melting Pt: 272 °C |
MDL Number:
MFCD00063060 CAS Number: 63-84-3 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...