1,8-Dihydroxy-9,10-anthraquinone ≥98.0% (by titrimetric analysis)
Supplier: TCI America
Synonyms:
Dantron, Danthron, 1,8-Dihydroxyanthraquinone
D0563-25G
TCD0563-25GEA
156
USD
TCD0563-25G
1,8-Dihydroxy-9,10-anthraquinone ≥98.0% (by titrimetric analysis)
1,8-Dihydroxy-9,10-anthraquinone
CAS Number: 117-10-2
MDL Number: MFCD00001211
Molecular Formula: C14H8O4
Molecular Weight: 240.21
Purity/Analysis Method:
Product Categories: Bioscience, Crude Medicine Ingredients for Research and Experimental Use, Other Pharmaceutical Ingredients, Others (Phytochemicals)
Other Product Names / Synonyms: Danthron, 1,8-Dihydroxyanthraquinone, Istizine
Formula:
C₁₄H₈O₄ MW: 240.22 g/mol Melting Pt: 193 °C |
MDL Number:
MFCD00001211 CAS Number: 117-10-2 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...