Digitonin
Supplier: TCI America
Synonyms:
Digitonin
D0540-100MG
TCD0540-100MGEA
104.91
USD
TCD0540-100MG
Digitonin
Digitonin
CAS Number: 11024-24-1
MDL Number: MFCD00077729
Molecular Formula: C56H92O29
Molecular Weight: 1229.32
Form: Crystal
Color: White
Melting point (°C): 240
Specific rotation [a]20/D: -52 deg (C=1, EtOH)
Formula:
C₅₆H₉₂O₂₉ MW: 1229.312 g/mol Melting Pt: 240 °C |
MDL Number:
MFCD00077729 CAS Number: 11024-24-1 UN: 3462 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...