N,N-Diethylformamide ≥99.0%
Supplier: TCI America
|
Warning
|
Synonyms:
Formic acid diethylamide, Formdiethylamide
D0506-500ML
D0506-25ML
D0506-100ML
TCD0506-500MLEA
301.09
USD
TCD0506-500ML
TCD0506-25ML
TCD0506-100ML
N,N-Diethylformamide ≥99.0%
N,N-Diethylformamide
CAS Number: 617-84-5
MDL Number: MFCD00003287
Molecular Formula: C5H11NO
Molecular Weight: 101.15
Purity/Analysis Method:
Other Product Names / Synonyms: Formyldiethylamine
Formula:
C₅H₁₁NO MW: 101.15 g/mol Boiling Pt: 176 Density: 0.91 (20 °C) Flash Pt: 60 °C Storage Temperature: Ambient |
MDL Number:
MFCD00003287 CAS Number: 617-84-5 EINECS: 210-533-2 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...