N,N-Diethylformamide ≥99.0%

Supplier: TCI America
Warning

Synonyms: Formic acid diethylamide, Formdiethylamide

D0506-500ML D0506-25ML D0506-100ML
TCD0506-500MLEA 301.09 USD
TCD0506-500ML TCD0506-25ML TCD0506-100ML
N,N-Diethylformamide ≥99.0%
N,N-Diethylformamide

CAS Number: 617-84-5 MDL Number: MFCD00003287 Molecular Formula: C5H11NO Molecular Weight: 101.15 Purity/Analysis Method: 99.0% (GC) Form: Clear Liquid Boiling point (°C): 177 Flash Point (°C): 60 Specific Gravity (20/20): 0.91


Other Product Names / Synonyms: Formyldiethylamine

Formula: C₅H₁₁NO
MW: 101.15 g/mol
Boiling Pt: 176
Density: 0.91 (20 °C)
Flash Pt: 60 °C
Storage Temperature: Ambient
MDL Number: MFCD00003287
CAS Number: 617-84-5
EINECS: 210-533-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4b585ec202c948282af306bb85baaddf12a0385d&vid=4b585ec202c948282af306bb85baaddf12a0385d&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR