Zinc diethyldithiocarbamate ≥99.0%

Supplier: TCI America

Synonyms: Zinc dethyldithiocarbamate, Diethyldithiocarbamic Acid Zinc Salt

D0492-500G D0492-25G
TCD0492-500GEA 183 USD
TCD0492-500G TCD0492-025G
Zinc diethyldithiocarbamate ≥99.0%
Zinc diethyldithiocarbamate

CAS Number: 14324-55-1 MDL Number: MFCD00064798 Molecular Formula: C5H11NS2 Molecular Weight: 361.90 Purity/Analysis Method: 99.0% (T) Form: Crystal Melting point (°C): 181 Flash Point (°C): 183


Other Product Names / Synonyms: Diethyldithiocarbamic Acid Zinc Salt

Formula: C₅H₁₁NS₂
MW: 361.90 g/mol
Melting Pt: 181 °C
Flash Pt: 183 °C
MDL Number: MFCD00064798
CAS Number: 14324-55-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9ed7fdeb7e7b8868db38cd959c44aa787b4ef8e6&vid=9ed7fdeb7e7b8868db38cd959c44aa787b4ef8e6&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR