1,3-Diethoxybenzene ≥97.0%

Supplier: TCI America

D0453-25G
TCD0453-025GEA 306 USD
TCD0453-025G
1,3-Diethoxybenzene ≥97.0%
1,3-Diethoxybenzene

CAS Number: 2049-73-2 MDL Number: MFCD00015145 Molecular Formula: C10H14O2 Molecular Weight: 166.22 Purity/Analysis Method: 97.0% (GC) Form: Clear Liquid Boiling point (°C): 235 Freezing point (°C): 10 Specific Gravity (20/20): 1.01


Other Product Names / Synonyms: Resorcinol Diethyl Ether

Formula: C₁₀H₁₄O₂
MW: 166.22 g/mol
Boiling Pt: 235 °C
Density: 1.01 (20 °C)
MDL Number: MFCD00015145
CAS Number: 2049-73-2
EINECS: 218-071-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=414acfeb16a58f1f792bf4fc09ad2ec340c94bd4&vid=414acfeb16a58f1f792bf4fc09ad2ec340c94bd4&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR