Ferrocene ≥98.0%
Supplier: TCI America
D0444-500G
D0444-25G
D0444-100G
TCD0444-500GEA
111.9
USD
TCD0444-500G
TCD0444-25G
TCD0444-100G
Ferrocene ≥98.0%
Ferrocene
CAS Number: 102-54-5
MDL Number: MFCD00001427
Molecular Formula: C10H10Fe
Molecular Weight: 186.04
Purity/Analysis Method:
Other Product Names / Synonyms: Bis(cyclopentadienyl)iron, Dicyclopentadienyl Iron
Formula:
C₁₀H₁₀Fe MW: 186.04 g/mol Boiling Pt: 249 Melting Pt: 176 °C Density: 1.49 g/cm³ (20 °C) Flash Pt: 33 °C |
MDL Number:
MFCD00001427 CAS Number: 102-54-5 EINECS: 203-039-3 UN: 1325 ADR: 4.1 II Merck Index: 13,04070 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...