Ferrocene ≥98.0%

Supplier: TCI America

D0444-500G D0444-25G D0444-100G
TCD0444-500GEA 111.9 USD
TCD0444-500G TCD0444-25G TCD0444-100G
Ferrocene ≥98.0%
Ferrocene

CAS Number: 102-54-5 MDL Number: MFCD00001427 Molecular Formula: C10H10Fe Molecular Weight: 186.04 Purity/Analysis Method: 98.0% (GC) Form: Crystal Boiling point (°C): 249 Melting point (°C): 176 Flash Point (°C): 33


Other Product Names / Synonyms: Bis(cyclopentadienyl)iron, Dicyclopentadienyl Iron

Formula: C₁₀H₁₀Fe
MW: 186.04 g/mol
Boiling Pt: 249
Melting Pt: 176 °C
Density: 1.49 g/cm³ (20 °C)
Flash Pt: 33 °C
MDL Number: MFCD00001427
CAS Number: 102-54-5
EINECS: 203-039-3
UN: 1325
ADR: 4.1 II
Merck Index: 13,04070
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c86c18cc123033b55408581546ac30c209c26c5e&vid=c86c18cc123033b55408581546ac30c209c26c5e&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR