Dichloro(methyl)silane ≥98.0% (by GC, titration analysis)

Supplier: TCI America

D0380-25G
TCD0380-25GEA 35.46 USD
TCD0380-25G
Dichloro(methyl)silane ≥98.0% (by GC, titration analysis)
Dichloro(methyl)silane

CAS Number: 75-54-7 MDL Number: MFCD00000494 Molecular Formula: CH4Cl2Si Molecular Weight: 115.03 Purity/Analysis Method: 98.0% (GC,T) Form: Clear Liquid Boiling point (°C): 41 Flash Point (°C): -20 Specific Gravity (20/20): 1.12 Storage Temperature: 0-10°C


Other Product Names / Synonyms: Methyldichlorosilane

Formula: CH₄Cl₂Si
Boiling Pt: 41 °C
Density: 1.12 (20 °C)
Flash Pt: –20 °C
MDL Number: MFCD00000494
CAS Number: 75-54-7
UN: 1242
ADR: 4.3,I
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b6486acdc62183dbdd2fae53ce2c28b789eb152f&vid=b6486acdc62183dbdd2fae53ce2c28b789eb152f&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR