3,5-Dichlorophenol ≥98.0%

Supplier: TCI America

D0365-10G
TCD0365-10GEA 82 USD
TCD0365-10G
3,5-Dichlorophenol ≥98.0%
3,5-Dichlorophenol

CAS Number: 591-35-5 MDL Number: MFCD00002259 Molecular Formula: C6H4Cl2O Molecular Weight: 163.00 Purity/Analysis Method: 98.0% (GC) Form: Crystal Boiling point (°C): 122 Melting point (°C): 68

Formula: C₆H₄Cl₂O
Boiling Pt: 122 °C
Melting Pt: 68 °C
MDL Number: MFCD00002259
CAS Number: 591-35-5
UN: 2020
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8cf8d46081de4187dba1aafb4ae55a234b1ef59a&vid=8cf8d46081de4187dba1aafb4ae55a234b1ef59a&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR