4-(Dimethylamino)stilbene ≥99.0% (by titrimetric analysis)
Supplier: TCI America
D0255-5G
TCD0255-005GEA
152
USD
TCD0255-005G
4-(Dimethylamino)stilbene ≥99.0% (by titrimetric analysis)
4-(Dimethylamino)stilbene
CAS Number: 1145-73-9
MDL Number: MFCD00017151
Molecular Formula: C16H17N
Molecular Weight: 223.32
Purity/Analysis Method:
Formula:
C₁₆H₁₇N Melting Pt: 149 °C |
MDL Number:
MFCD00017151 CAS Number: 1145-73-9 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...