Chromeazurol S (Mordant blue 29)
Supplier: TCI America
|
Warning
|
D0241-25G
TCD0241-025GEA
100.05
USD
TCD0241-025G
Chromeazurol S (Mordant blue 29)
Chromeazurol S (Mordant blue 29)
CAS Number: 1667-99-8
MDL Number: MFCD00001615
Molecular Formula: C23H16Cl2O9S
Molecular Weight: 605.28
Form: Crystal
Color: Yellow Red
Form: Crystal
Color: Yellow Red
Formula:
C₂₃H₁₆Cl₂O₉S |
MDL Number:
MFCD00001615 CAS Number: 1667-99-8 |
|
Specification Test Results
Lambda max. | 425.0 to 435.0 nm(pH7.0 Buffer sol.) |
Absorbance | min. 0.300(0.02g/L, pH7.0 Buffer sol., 425.0 to 435.0 nm)(calcd.on dried substance) |
Drying loss | max. 10.0 % |
Sensitiveness | Colorimetric Test : Turn Purple with Aluminum and turn Yellow red by adding EDTA 2Na |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...