Dibutyl disulfide ≥95.0%

Supplier: TCI America
Warning

Synonyms: Dibutyldisulphide, n-Butyl disulfide, n-Butyl disulphide, Dibutyldisulfide

D0224-25ML D0224-500ML
TCD0224-025MLEA 49 USD
TCD0224-025ML TCD0224-500ML
Dibutyl disulfide ≥95.0%
Dibutyl disulphide

CAS Number: 629-45-8 MDL Number: MFCD00009467 Molecular Formula: C8H18S2 Molecular Weight: 178.35 Purity/Analysis Method: 95.0% (GC) Form: Clear Liquid Boiling point (°C): 88 Flash Point (°C): 93 Specific Gravity (20/20): 0.94


Other Product Names / Synonyms: Butyl Disulfide

Formula: C₈H₁₈S₂
MW: 178.36 g/mol
Boiling Pt: 88 °C
Melting Pt: –94 °C
Density: 0.94 (20 °C)
Flash Pt: 93 °C
Storage Temperature: Ambient
MDL Number: MFCD00009467
CAS Number: 629-45-8
EINECS: 211-091-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=50bd5f84aa43cacace3d8ba3a8fd653cb611cd62&vid=50bd5f84aa43cacace3d8ba3a8fd653cb611cd62&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR