1,3-Dibromobenzene ≥97.0%

Supplier: TCI America
Warning

D0169-25G D0169-500G D0169-100G
TCD0169-025GEA 25 USD
TCD0169-025G TCD0169-500G 76218-950
1,3-Dibromobenzene ≥97.0%
1,3-Dibromobenzene

CAS Number: 108-36-1 MDL Number: MFCD00000078 Molecular Formula: C6H4Br2 Molecular Weight: 235.91 Purity/Analysis Method: 97.0% (GC) Form: Clear Liquid Boiling point (°C): 219 Melting point (°C): -7 Flash Point (°C): 94 Specific Gravity (20/20): 1.96

Formula: C₆H₄Br₂
Boiling Pt: 219 °C
Melting Pt: –7 °C
Density: 1.96 (20 °C)
Flash Pt: 94 °C
Storage Temperature: Ambient
MDL Number: MFCD00000078
CAS Number: 108-36-1
EINECS: 203-574-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=cedf774ec1fa5c877b6939b2ffe0447b0a9f2f74&vid=3946573d879897b33d8463c885bcdbf07b773358&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR