Dibenz[a,h]anthracene ≥98.0%

Supplier: TCI America
Danger

Synonyms: Naphtho[1',2':2,3]phenanthrene

D0145-100MG
TCD0145-100MGEA 78 USD
TCD0145-100MG
Dibenz[a,h]anthracene ≥98.0%
Dibenz[a,h]anthracene

CAS Number: 53-70-3 MDL Number: MFCD00003708 Molecular Formula: C22H14 Molecular Weight: 278.35 Purity/Analysis Method: 98.0% (GC) Form: Crystal Boiling point (°C): 524 Melting point (°C): 266


Product Categories: Materials Chemistry, Nanocarbon Unit Structures (Nanocarbon Materials) Other Product Names / Synonyms: Naphtho[1',2':2,3]phenanthrene

Formula: C₂₂H₁₄
MW: 278.35 g/mol
Boiling Pt: 524
Melting Pt: 266
Density: 1.282 g/cm³ (20 °C)
MDL Number: MFCD00003708
CAS Number: 53-70-3
EINECS: 200-181-8
UN: 3077
ADR: 9 III
Merck Index: 12,03054
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d4329002ab5c65adeac1bdfcc1593453cb3c7e1f&vid=d4329002ab5c65adeac1bdfcc1593453cb3c7e1f&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR