3,4-Diaminotoluene ≥98.0% (by titrimetric analysis)

Supplier: TCI America

D0125-25G
TCD0125-025GEA 89 USD
TCD0125-025G
3,4-Diaminotoluene ≥98.0% (by titrimetric analysis)
3,4-Diaminotoluene

CAS Number: 496-72-0 MDL Number: MFCD00007728 Molecular Formula: C7H10N2 Molecular Weight: 122.17 Purity/Analysis Method: 98.0% (T) Form: Crystal Boiling point (°C): 265 Melting point (°C): 90


Other Product Names / Synonyms: 3,4-Toluenediamine, Tolylene-3,4-diamine

Formula: C₇H₁₀N₂
Boiling Pt: 265 °C
Melting Pt: 90 °C
Storage Temperature: Ambient
MDL Number: MFCD00007728
CAS Number: 496-72-0
EINECS: 207-826-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8d71aa3fc4152d3240889071c6c135004b1dc1c5&vid=8d71aa3fc4152d3240889071c6c135004b1dc1c5&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR