1,3-Dithiane ≥97.0%

Supplier: TCI America

D0119-25G D0119-5G
TCD0119-25GEA 267.67 USD
TCD0119-25G TCD0119-5G
1,3-Dithiane ≥97.0%
1,3-Dithiane

CAS Number: 505-23-7 MDL Number: MFCD00006654 Molecular Formula: C4H8S2 Molecular Weight: 120.23 Purity/Analysis Method: 97.0% (GC) Form: Crystal Boiling point (°C): 208 Melting point (°C): 53

Formula: C₄H₈S₂
Boiling Pt: 208 °C
Melting Pt: 53 °C
MDL Number: MFCD00006654
CAS Number: 505-23-7
UN: 3335
ADR: 9,
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4f06af767faa04fa2786a9480b493cbad32a253a&vid=4f06af767faa04fa2786a9480b493cbad32a253a&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR