[8]Cycloparaphenylene ≥95.0% (by HPLC)

Supplier: TCI America

Synonyms: [8]CPP

C3544-20MG
TCC3544-20MGEA 501 USD
TCC3544-20MG
[8]Cycloparaphenylene ≥95.0% (by HPLC)
[8]Cycloparaphenylene
Formula: C₄₈H₃₂
MW: 608.78 g/mol
Melting Pt: 300 °C
CAS Number: 1217269-85-6

Order Now

Specification Test Results

Appearance White to Yellow to Green powder to crystal
Purity (HPLC) min. 95.0 area%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR