trans-Cinnamic acid ≥99.5% (by GC, titration analysis) matrix for MALDI-TOF/MS

Supplier: TCI America
Warning

C3412-1G
TCC3412-1GEA 53 USD
TCC3412-1G
trans-Cinnamic acid ≥99.5% (by GC, titration analysis) matrix for MALDI-TOF/MS
trans-Cinnamic acid
Formula: C₉H₈O₂
Boiling Pt: 300 °C
MDL Number: MFCD00004369
CAS Number: 140-10-3
EINECS: 205-398-1
Merck Index: 14,02299

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 99.5 %
Purity (Neutralization titration) min. 99.5 %
Melting point 132.0 to 136.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR