3-Cyano-2-fluorophenylboronic acid (contains varying amounts of Anhydride)

Supplier: TCI America
Warning

C3214-1G
TCC3214-1GEA 99 USD
TCC3214-1G
3-Cyano-2-fluorophenylboronic acid (contains varying amounts of Anhydride)
3-Cyano-2-fluorophenylboronic acid
Formula: C₇H₅BFNO₂
MDL Number: MFCD07374984
CAS Number: 957121-05-0

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (Neutralization titration) 98.0 to 112.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR