4-Cyanophenyl isothiocyanate ≥98.0% (by GC)

Supplier: TCI America
Danger

Synonyms: 4-Cyanophenylisothiocyanate, 1-Cyano-4-isothiocyanatobenzene, 4-Isothiocyanatobenzonitrile

C3169-5G
TCC3169-5GEA 154 USD
TCC3169-5G
4-Cyanophenyl isothiocyanate ≥98.0% (by GC)
4-Cyanophenyl isothiocyanate
Formula: C₈H₄N₂S
MW: 160.2
Boiling Pt: 294
Storage Temperature: Refrigerator
MDL Number: MFCD00041085
CAS Number: 2719-32-6
EINECS: 220-323-2
UN: 2811
ADR: 6.1 III

Order Now

Specification Test Results

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 98.0 %
Melting point 119.0 to 123.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR