Carbon nanotube single-walled, Length ∼5 µm, Diameter <∼3nm (>85%)

Supplier: TCI America
Warning

Synonyms: CNT

C3133-200MG
TCC3133-200MGEA 203.51 USD
TCC3133-200MG
Carbon nanotube single-walled, Length ∼5 µm, Diameter <∼3nm (>85%)
Carbon nanotube
Formula: C
MDL Number: MFCD00133992
CAS Number: 308068-56-6

Order Now

Specification Test Results

Appearance Black powder to crystal
Purity (TPO Method) min. 85 %
Diameter max. 3 nm
Length min. 5 micro_m


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR