2-chloro-5-iodopyrimidine ≥98.0% (by GC)

Supplier: TCI America
Warning

Synonyms: 2-Chloro-5-iodopyrimidine

C3119-1G
TCC3119-1GEA 98 USD
TCC3119-1G
2-chloro-5-iodopyrimidine ≥98.0% (by GC)
2-chloro-5-iodopyrimidine
Formula: C₄H₂ClIN₂
MDL Number: MFCD06200210
CAS Number: 32779-38-7

Order Now

Specification Test Results

Appearance White to Gray to Brown powder to crystal
Purity (GC) min. 98.0 %
Melting point 133.0 to 137.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR