Cerium triacetate hydrate ≥98.0% (by titrimetric analysis)

Supplier: TCI America
Warning

C2974-25G C2974-500G
TCC2974-25GEA 55 USD
TCC2974-25G TCC2974-500G
Cerium triacetate hydrate ≥98.0% (by titrimetric analysis)
Cerium triacetate hydrate
Formula: C₆H₉CeO₆·H₂O
Melting Pt: 308°C
MDL Number: MFCD00150533
CAS Number: 206996-60-3
EINECS: 208-654-0

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (Chelometric Titration) min. 98.0 % (calcd.on anh.substance)
Water 4.0 to 10.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR