2-Chloroimidazole ≥98.0%

Supplier: TCI America

Synonyms: 2-Chloro-1H-imidazole

C2685-1G
TCC2685-1GEA 286 USD
TCC2685-1G TCC2685-200MG
2-Chloroimidazole ≥98.0%
2-Chloroimidazole

CAS Number: 16265-04-6 MDL Number: MFCD02179530 Molecular Formula: C3H3ClN2 Molecular Weight: 102.52 Purity/Analysis Method: 98.0% (GC) Form: Crystal Melting point (°C): 168 Storage Temperature: 0-10°C

Formula: C₃H₃ClN₂
MW: 102.52
Melting Pt: 168 °C
MDL Number: MFCD02179530
CAS Number: 16265-04-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=bd4ac0b18181206dbc34b7148210a702e79c1f9e&vid=bd4ac0b18181206dbc34b7148210a702e79c1f9e&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR