6-Chloro-m-toluidine ≥98.0%

Supplier: TCI America

C2576-5G
TCC2576-5GEA 61.7 USD
TCC2576-5G
6-Chloro-m-toluidine ≥98.0%
6-Chloro-m-toluidine

CAS Number: 95-81-8 MDL Number: MFCD00007674 Molecular Formula: C7H8ClN Molecular Weight: 141.60 Purity/Analysis Method: 98.0% (GC,T) Form: Crystal Boiling point (°C): 230 Flash Point (°C): 107 Freezing point (°C): 29


Other Product Names / Synonyms: 3-Amino-4-chlorotoluene, 6-Chloro-m-toluidine

Formula: C₇H₈ClN
Boiling Pt: 230 °C
Flash Pt: 107 °C
MDL Number: MFCD00007674
CAS Number: 95-81-8
UN: 2239
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=34f4fb44873781ba07d8f0b747e28382a3c6523b&vid=34f4fb44873781ba07d8f0b747e28382a3c6523b&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR