2-Chloro-3-pyridineboronic acid (contains varying amounts of Anhydride)
Supplier: TCI America
CAS Number: 381248-04-0
MDL Number: MFCD03094997
Molecular Formula: C5H5BClNO2
Molecular Weight: 157.36
Form: Crystal
Color: White
Melting point (°C): 160
Other Product Names / Synonyms: 2-Chloro-3-pyridylboronic Acid
Formula:
C₅H₅BClNO₂ Melting Pt: 160 °C |
MDL Number:
MFCD03094997 CAS Number: 381248-04-0 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...